Learn More
Invitrogen™ ADAM21 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ADAM21 protein.
Specifications
Specifications
| Antigen | ADAM21 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ADAM21 |
| Gene Accession No. | Q9UKJ8 |
| Gene Alias | a disintegrin and metallopeptidase domain 21; a disintegrin and metalloprotease domain (ADAM) 21; a disintegrin and metalloprotease domain 21; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 21; ADAM; ADAM 21; ADAM 31; ADAM metallopeptidase domain 21; ADAM metallopeptidase domain 21, preproprotein; Adam21; Adam31; ADAMs; disintegrin and metalloproteinase domain-containing protein 21; Disintegrin and metalloproteinase domain-containing protein 31; metalloendopeptidases |
| Gene Symbols | ADAM21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.