Learn More
Invitrogen™ ADAM8 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Specifications
Specifications
| Antigen | ADAM8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ADAM8 |
| Gene Accession No. | P78325 |
| Gene Alias | a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394 |
| Gene Symbols | ADAM8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.