Learn More
Description
- ADAM9 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human ADAM9.
Specifications
Specifications
| Description | ADAM metallopeptidase domain 9 (meltrin gamma) |
| Cross Reactivity | Human |
| Product Type | Kit |
| Gene ID (Entrez) | 8754 |
| Gene Symbol | ADAM9 |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.