Learn More
Sigma Organic Chemistry Adamantane-1-carboxamiD | 100mg | MFCD00167793
Supplier: Sigma Organic Chemistry CDS002865100MG

Adamantane-1-carboxamiD , 100mg
About this Item:
MDL #: MFCD00167793
Molecular Weight: 214.73
UNSPSC Code: 12352200
Chemical Formula: C11H19ClN2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.