Learn More
Adamantane, 98%
Description
Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 266°C to 268°C (sealed tube) |
| Density | 1.07 |
| Quantity | 1000 g |
| Beilstein | 1901173 |
| Merck Index | 14,149 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 136.24 |
| Percent Purity | 98% |
| Chemical Name or Material | Adamantane |
Safety and Handling
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 206-001-4 |
| RTECSNumber | YK0493000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.