Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMDEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526128
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526128 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526128 Supplier Invitrogen™ Supplier No. PA526128
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMDEC1 Polyclonal Antibody for Western Blot, IHC (P), Flow

ADAMDEC1 may play an important role in the control of the immune response and during pregnancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMDEC1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADAMDEC1
Gene Accession No. O15204
Gene Alias 2210414L24Rik; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain-like protein decysin 1; a disintegrin and metalloproteinase domain-like protein decysin-1; ADAM; ADAM DEC1; ADAM like decysin 1; ADAMDEC1; ADAM-like protein decysin 1; ADAM-like protein decysin-1; ADAM-like, decysin 1; ADAMs; AI574231; Dcsn; decysin; disintegrin metalloprotease (decysin); disintegrin protease; M12.219; metalloendopeptidases; Unknown (protein for IMAGE:7929269)
Gene Symbols ADAMDEC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 39-69 amino acids from the N-terminal region of human ADAMDEC1.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.