Learn More
Description
Recommended positive controls: HCT116, MCF-7, JC. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ADAMTS5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ADAMTS5 |
| Gene Accession No. | Q9R001, Q9UNA0 |
| Gene Alias | 9530092O11Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 5; ADAM metallopeptidase with thrombospondin type 1 motif, 5; ADAMs; ADAM-TS 11; ADAM-TS 5; ADAMTS family member; ADAMTS1; ADAMTS11; ADAMTS-11; ADAMTS5; ADAM-TS5; ADAMTS-5; ADMP2; ADMP-2; Aggrecanase 2; Aggrecanase-2; AI481094; ASMP-2; Implantin; metalloendopeptidases |
| Gene Symbols | ADAMTS5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
