Learn More
Description
PA1-1742 detects ADAMTS5 in human samples. PA1-1742 has been successfully used in Western blot procedures. In Western blot analysis of JJ012 cells this antibody detects a ~64 kDa protein representing ADAMTS5. The PA1-1742 immunogen is a synthetic peptide corresponding to residues C I(442) L T S I D A S K P(451) of human ADAMTS5.
Specifications
Specifications
| Antigen | ADAMTS5 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | ADAMTS5 |
| Gene Accession No. | Q9UNA0 |
| Gene Alias | 9530092O11Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 5; ADAM metallopeptidase with thrombospondin type 1 motif, 5; ADAMs; ADAM-TS 11; ADAM-TS 5; ADAMTS family member; ADAMTS1; ADAMTS11; ADAMTS-11; ADAMTS5; ADAM-TS5; ADAMTS-5; ADMP2; ADMP-2; Aggrecanase 2; Aggrecanase-2; AI481094; ASMP-2; Implantin; metalloendopeptidases |
| Gene Symbols | ADAMTS5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
