Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTSL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596920 Supplier Invitrogen™ Supplier No. PA596920
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLVIQDYWWS VDRLATCSAS CGNRGVQQPR LRCLLNSTEV NPAHCAGKVR PAVQPIACNR RDCPSRWMVT SWSACTRSCG GGVQTRRVTC QKLKASGIST PVSNDMCTQV AKRPVDTQAC NQQLCVEWAF SSWGQCNGPC IGPHLAVQHR QVFCQTRDGI TLPSEQCSAL PRPVSTQNCW SEACSVHWRV SLWTLCTATC GNYGFQSRRV ECVHARTNKA VPEHLCSWGP RPANWQRCNI TPCENMECRD TTRYCEKVKQ LKLCQLSQFK SRCCGTCGKA; Positive Samples: U-87MG, A-375, HeLa, Mouse heart, Mouse spleen, Rat heart; Cellular Location: extracellular matrix, extracellular space.

Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADAMTSL1
Gene Accession No. Q8BLI0, Q8N6G6
Gene Alias 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071
Gene Symbols ADAMTSL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1483-1762 of human ADAMTSL1 (NP_0010353623).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362539, 77739, 92949
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.