Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Adapalene

Catalog No. 285210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-5210 10 mg
28-525-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-5210 Supplier Tocris Bioscience™ Supplier No. 2852/10
This item is not returnable. View return policy

RARβ and RARγ agonist

Retinoic acid analog that is a RARβ and RARγ agonist (AC50 values are 2.2, 9.3, 22 and > 1000 nM for RARβ, RARγ, RARα and RXRα receptors respectively). Inhibits proliferation and induces apoptosis in colorectal cancer cell in vitro. Displays comedolytic activity.

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3

Specifications

Quantity 10 mg
Formula Weight 412.52
Purity >98%
Molecular Weight (g/mol) 412.529
Chemical Name or Material Adapalene
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.