Learn More
Research Products International Corp Strip Spin Adapter for 0.2ML tubes for Standard Spectrafuge Rotor
SDP
Supplier:  Research Products International Corp 141307TSTR
Encompass_Preferred
StripSpin adapter for use with Spectrafuge 24D for 0.2 ml tubes and strips.Shipping Weight: 1 lbsShipping Dimensions: 4.00 x 4.00 x 4.00Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-222-9732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.