Learn More
Beckman Coulter FisherbrandTM Disposable Polypropylene Beakers, 100 mL, Sterile, 500/Pack
Supplier:  Beckman Coulter 359149
Encompass_Preferred
The FisherbrandTM NC2198887 is a polypropylene microcentrifuge tube rack that holds 96 1.5mL tubes in an 8x12 array. It is made of autoclavable polypropylene and has alphanumeric coordinates to identify tube positions. The rack measures 12.7cm x 8.6cm x 5.4cm. This product can be used to hold microcentrifuge tubes upright and organized during storage and processing. The sturdy construction allows it to be reused. It is an essential labware accessory for any molecular biology or biochemistry lab working with 1.5mL microcentrifuge tubes.
Catalog No. NC2198887
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.