Learn More
Eppendorf™ Adapter, for 12 tubes 50 mL Nalgene™ Oak Ridge, 2 pcs.
Catalog No. 05414362 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Rotor S-4 xUniversal-Large
Catalog No. For Use With (Equipment)
05-414-362 Rotor S-4 xUniversal-Large
1 options
This item is not returnable. View return policy
Catalog No. 05-414-362 Supplier Eppendorf™ Supplier No. 5920753002
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter, for 12 tubes 50 mL Nalgene™ Oak Ridge, for Rotor S-4xUniversal-Large, 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapters, Buckets, and Accessories
For Use With (Equipment) Rotor S-4 xUniversal-Large
Vessels per Adapter 12
No. per Pack 2