Learn More
Chemglass Life Sciences Adapter, 1" Beaded Pipe to 1" Sanitary, GFTFE
SDP
Supplier:  Chemglass Life Sciences CG1964B12
Encompass_Preferred
Adapter, 1" Beaded Pipe to 1" Sanitary, GFTFE. Beaded pipe to sanitary fitting adapter connects to the 1¿ beaded pipe side arm of CG-1968-Q zero dead space drain valve. Approximate overall length of adapter is 2¿(52mm). NOTE: Requires a CG-1968-66 beaded coupling.
Catalog No. 50-131-1165