Learn More
Eppendorf™ Adapter for 25/50 mL Conical Tubes and Plates
Catalog No. 05413978 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
25 mL/50 mL conical tubes and plates
Catalog No. For Use With (Equipment)
05-413-978 25 mL/50 mL conical tubes and plates
1 options
Catalog No. 05-413-978 Supplier Eppendorf™ Supplier No. 5920756001
Add to Cart
Add to Cart
Conical tube and plate adapter for rotor S-4xUniversal-Large.
  • 2 pieces
  • Adapter
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
For Use With (Equipment) 25 mL/50 mL conical tubes and plates
Description For Rotor S-4xUniversal-Large
No. per Pack 2