Learn More
Chemglass Life Sciences Adapter, Outer to 29/26 Inner, Vacuum, Stopcock
SDP
Supplier:  Chemglass Life Sciences CG1049J29
Encompass_Preferred
Adapter, Outer to 29/26 Inner, Vacuum, Stopcock. Vacuum Adapter has a ground glass standard taper outer joint on top that provides a convenient leak free way to connect to the OP-6602, OP-6603 and OP-6604 disposable long stem filter funnels and a taper inner joint or a vial. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. The 2mm PTFE stopcock between the outer joint and standard taper inner permits control of the vacuum source. Hose connection is 10mm OD at the largest serration. U.S. patent pending.
Catalog No. 50-121-4823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.