Learn More
Eppendorf™ Adapter
Catalog No. 05414134 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-44
Catalog No. For Use With (Equipment)
05-414-134 Rotor A-4-44
1 options
This item is not returnable. View return policy
Catalog No. 05-414-134 Supplier Eppendorf™ Supplier No. 022637479
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter for use with rectangular bucket in Rotor A-4-44.
  • For 1 conical tubes 50 mL
  • For 100 mL rectangular bucket in Rotor A-4-44
  • 8 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical
Max. Tube Length 50 mL
For Use With (Equipment) Rotor A-4-44
Vessels per Adapter 1