Learn More
Eppendorf™ Adapter
Catalog No. 05414152 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor S-4x500
Catalog No. For Use With (Equipment)
05-414-152 Rotor A-4-81 and Rotor S-4x500
1 options
This item is not returnable. View return policy
Catalog No. 05-414-152 Supplier Eppendorf™ Supplier No. 022638904
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter for Rotor A-4-81 and Rotor S-4x500.
  • For 5 tubes 30 to 50 mL round-bottom/50 mL conical, skirted.
  • Please order adapter for 50 mL conical tubes skirted.
  • For 500 mL rectangular bucket in Rotor A-4-81 and Rotor S-4x500
  • 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical, Skirted
For Use With (Equipment) Rotor A-4-81 and Rotor S-4x500
Vessels per Adapter 5
Tube Size 30 to 50 mL