Learn More
Eppendorf™ Adapter
Catalog No. 05414153 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor S-4x500
Catalog No. For Use With (Equipment)
05-414-153 Rotor A-4-81 and Rotor S-4x500
1 options
This item is not returnable. View return policy
Catalog No. 05-414-153 Supplier Eppendorf™ Supplier No. 022638921
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter for Rotor A-4-81 and Rotor S-4x500.
  • For 1 bottle 180 to 250 mL flat/175 to 225 mL conical.
  • For 500 mL rectangular bucket in Rotor A-4-81 and Rotor S-4x500.
  • Please note that these conical bottles need to be used together with a conical form insert provided by the manufacturer.
  • 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Flat/Conical Bottom
For Use With (Equipment) Rotor A-4-81 and Rotor S-4x500
Tube Size 175 to 250 mL