Learn More
Eppendorf™ Adapter
Catalog No. 05414164 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
1 x 15 mL Tube
Catalog No. Capacity
05-414-164 1 x 15 mL Tube
1 options
This item is not returnable. View return policy
Catalog No. 05-414-164 Supplier Eppendorf™ Supplier No. 022639447
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter for 1 conical tube 15 mL, for Rotor F-35-30-17.
  • For 1 conical tube 15 mL
  • For Rotor F-35-30-17
  • 10 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical
Capacity 1 x 15 mL Tube
For Use With (Equipment) Rotor F-35-30-17