Learn More
ITL BioMedical Adapter Cap with Female Luer
Catalog No. 23900124 Shop All ITL BioMedical Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
23-900-124 Single-use
1 options
This item is not returnable. View return policy
Catalog No. 23-900-124 Supplier ITL BioMedical Supplier No. ITL 10002
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Adapter Cap with Female Luer offers a safe, efficient method for transferring blood samples from syringes into blood culture bottles or blood collection tubes. Designed to streamline sample collection and reduce contamination risks, this adapter cap is an essential tool for clinical settings prioritizing safety and efficiency.
  • Eliminates the need for additional needles, reducing contamination risks and exposure to sharps.
  • Effortlessly connects to blood culture bottles or blood collection tubes for streamlined sampling.
  • Enables secure, direct blood transfer from syringe to blood culture bottles or blood collection tubes, preserving sample quality.
  • Designed for quick detachment and safe, compliant disposal after use.
Specifications
Product Type Adapter Cap
Autoclavable Not Autoclavable
Material Polypropylene
Color Blue
Sterility Sterile
Disposable Single-use
Fit for Purpose Sterile