Learn More
ITL BioMedical Adapter Cap with Male Luer
Catalog No. 22515359 Shop All ITL BioMedical Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
22-515-359 Single-use
1 options
This item is not returnable. View return policy
Catalog No. 22-515-359 Supplier ITL BioMedical Supplier No. ITL 10001
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Adapter Cap with Male Luer Lock enables direct connection to blood collection winged needles and peripheral lines, facilitating a safe and effective blood culture collection process. Engineered to ensure precise sample handling and reduce the risks associated with blood collection, this adapter cap is ideal for labs, clinics, and medical offices.
  • Secure male luer lock provides a stable, needle-free connection to blood collection sets.
  • Compatible with blood culture bottles and blood collection tubes, offering flexibility in sample collection.
  • Designed to maintain sample integrity and ensure efficient blood transfer.
  • Easily removable for quick, safe disposal according to facility protocols.
Specifications
Product Type Adapter Cap
Autoclavable Not Autoclavable
Material Polypropylene
Color Blue
Sterility Sterile
Disposable Single-use
Fit for Purpose Sterile