Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Adapter for Centrifuge 5910 R with Rotor S-4xUniversal
Shop All Eppendorf North America ProductsDescription
- For 1,000 mL wide-neck bottle (Catalog No. 5910 770.006) in combination with Rotor S-4xUniversal
- Set of 2 pieces
Specifications
Specifications
| Capacity | 1 x 1000 mL |
| Description | For Centrifuge 5910 R with Rotor S-4xUniversal |
| No. of Places | 1 |
| No. per Pack | Set of 2 pieces |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?