Learn More
Eppendorf™ Adapter for Conical Tubes, Bottles, and Plates
Catalog No. 05413979 Shop All Eppendorf North America Products
Change view
Click to view available options
No. per Pack:
2
Catalog No. No. per Pack
05-413-979 2
1 options
Catalog No. 05-413-979 Supplier Eppendorf™ Supplier No. 5920755005
Add to Cart
Add to Cart
Conical tube, bottle, and plate adapter for rotor S-4xUniversal-Large.
  • 2 pieces
  • Adapter
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
For Use With (Equipment) 50 mL conical tubes, 175 to 250 mL bottles, and plates
Description For Rotor S-4xUniversal-Large
No. per Pack 2