Learn More
Electron Microscopy Sciences Evactron Adapter Flange HITACHI, 69 mm, 47.5 mm O-ring ID

Supplier: Electron Microscopy Sciences 23056801

Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 69 mm
O-ring ID: 47.5 mm
Mounting holes: 4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.