Learn More
Electron Microscopy Sciences Evactron Adapter Flange JEOL, 154x204 mm
SDP
Supplier:  Electron Microscopy Sciences 23032501
Encompass_Preferred
Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 154 x 204 mm
Mounting holes: 4 ASYM
Catalog No. 50-192-8259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.