Learn More
Electron Microscopy Sciences Evactron Adapter Flange ZEISS, 68x88 mm, OVAL O-RING
SDP
Supplier:  Electron Microscopy Sciences 23003801
Encompass_Preferred
Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 68 x 88 mm
O-ring ID: OVAL O-RING
Mounting holes: 4 SYMM
Catalog No. 50-192-8272