Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Adapter, Joint to Serrated Hose
GREENER_CHOICESDP

Catalog No. 31501809 Shop All Kemtech Products
Change view
Click to view available options
Fits Tubing ID (Metric):
10 mm
12 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fits Tubing ID (Metric)
31-501-809 10 mm
31-501-810 12 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-809 Supplier Synthware™ Supplier No. M131604
Only null left
Add to Cart
Add to Cart

Adapter, No. 15 O-ring joint with serrated hose connection

Specifications

For Use With (Application) Organic Synthesis
Fits Tubing ID (Metric) 10 mm
Material Borosilicate Glass
Product Type #15 O-Ring Joint to 10 mm Serrated Hose Connection
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.