Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Adapter for Rotors S-4-104, S-4x750, S-4x1000

Catalog No. 05414343 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
8 x 30 mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
05-414-343 8 x 30 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-414-343 Supplier Eppendorf™ Supplier No. 5825755006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitates use of 30 mL conical tube array in rotor buckets for S-4-104, S-4x750, S-4x1000 rotors.

  • Suitable for S-4-104, S-4x750, S-4x1000 rotor buckets
  • Accepts 8 x 30 mL conical tubes
TRUSTED_SUSTAINABILITY

Specifications

Tube Type Conical
Capacity 8 x 30 mL Tubes
For Use With (Equipment) S-4-104, S-4x750, S-4x1000 rotors and buckets
Vessels per Adapter 8
No. of Places 8
No. per Pack 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.