Learn More
Ace Glass, Inc. Thermocouple adapter, 24/29, use with 6.35mm diameter probes, complete with PTFE bushing and FETFE; O-rings
SDP
Supplier:  Ace Glass, Inc. 5041124
Encompass_Preferred
Ace-Thred to 14/23 standard taper Thermocouple adapter with a PTFE bushing and PTFE ferrule. Thermocouple adapter is designed specifically to hold a 3.175mm diameter temperature probe, thermometer, or tubing.
Catalog No. 50-971-682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.