Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bottle Top Dispenser Adapter

Catalog No. 13681522 Shop All Fisherbrand Products
Change view
Click to view available options
Model:
ADPT-36
ADPT-38, -40, -45
ADPT-40
ADPT-45
Diameter (Metric):
36 mm
38, 40, 45 mm
40 mm
45 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Diameter (Metric)
13-681-522 ADPT-45 45 mm
13-681-520 ADPT-36 36 mm
13-681-523 ADPT-38, -40, -45 38, 40, 45 mm
13-681-521 ADPT-40 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 13-681-522
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured with PP material and can withstand fumic reagents. This Bottle Top Dispenser Adapter is durable and light weight.

  • High durable Quality
  • Manufactured with PP material, hence can withstand fumic reagents
  • Very light weighted
ADPT-45 Inlet/Outlet Size: 
  • Inlet Dia: 45 mm (without thread); 42.4 mm (with thread)
  • Outlet Dia: 29.4 mm (without thread); 32 mm (with thread)

Warranty not applicable for spare parts

Specifications

Product Type Adapter
Certifications/Compliance CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025, Above certificates applicable for the whole product
Color Translucent
Material Polypropylene
For Use With (Equipment) Bottletop Dispenser
Model ADPT-45
Shape Round
Diameter (Metric) 45 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.