Learn More
Thermo Scientific™ Adapters for TX-150 Swinging Bucket Rotors
GSA_VA
Catalog No. 75005739
Change view
Click to view available options
Tube Type:
100 mL Round Bottom Open-Top Tube
11 mL IVF Tube
13 mL Urine Tube
25 mL Universal Flat/Skirted Tube
3/5 mL Blood Collection Tube
5/7 mL Blood Collection Tube
50 mL Conical or Skirted Tube
Microtube
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Type
75-005-739 5/7 mL Blood Collection Tube
75-005-744 25 mL Universal Flat/Skirted Tube
75-005-740 3/5 mL Blood Collection Tube
75-005-737 11 mL IVF Tube
75-005-736 50 mL Conical or Skirted Tube
75-005-735 100 mL Round Bottom Open-Top Tube
75-005-743 Microtube
75-003-504 13 mL Urine Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 75-005-739 Supplier Thermo Scientific™ Supplier No. 75005739
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assorted tube adapters
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type 5/7 mL Blood Collection Tube
Capacity 5/7 mL
Max. Tube Diameter 13 mm
Max. Tube Length 116 mm
For Use With (Equipment) 50mL Conical Buckets for TX-150 rotor