Learn More
Electron Microscopy Sciences Evactron Adapter Flange ZEISS, 150x190 mm
SDP
Supplier:  Electron Microscopy Sciences 23036601
Encompass_Preferred
Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 150 x 190 mm
Mounting holes: 10 ASYM
Catalog No. 50-192-8278