Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAR Monoclonal Antibody (GT1066)
GREENER_CHOICE

Catalog No. PIMA517285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517285 Supplier Invitrogen™ Supplier No. MA517285
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, A375. Predicted reactivity: Mouse (80%), Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Adenosine Deaminase RNA Specific (ADAR) catalyzes the hydrolytic deamination of adenosine to inosine in double-stranded RNA (dsRNA) referred to as A-to-I RNA editing.This may affect gene expression and function in a number of ways that include mRNA translation by changing codons and hence the amino acid sequence of proteins; pre-mRNA splicing by altering splice site recognition sequences; RNA stability by changing sequences involved in nuclease recognition; genetic stability in the case of RNA virus genomes by changing sequences during viral RNA replication; and RNA structure-dependent activities such as microRNA production or targeting or protein-RNA interactions. ADAR can edit both viral and cellular RNAs and can edit RNAs at multiple sites (hyper-editing) or at specific sites (site-specific editing). Its cellular RNA substrates include: bladder cancer-associated protein (BLCAP), neurotransmitter receptors for glutamate (GRIA2) and serotonin (HTR2C) and GABA receptor (GABRA3). Site-specific RNA editing of transcripts encoding these proteins results in amino acid substitutions which consequently alters their functional activities. Exhibits low-level editing at the GRIA2 Q/R site, but edits efficiently at the R/G site and HOTSPOT1. Its viral RNA substrates include: hepatitis C virus (HCV), vesicular stomatitis virus (VSV), measles virus (MV), hepatitis delta virus (HDV), and human immunodeficiency virus type 1 (HIV-1). Exhibits either a proviral (HDV, MV, VSV and HIV-1) or an antiviral effect (HCV) and this can be editing-dependent (HDV and HCV), editing-independent (VSV and MV) or both (HIV-1). Impairs HCV replication via RNA editing at multiple sites. Enhances the replication of MV, VSV and HIV-1 through an editing-independent mechanism via suppression of EIF2AK2/PKR activation and function. Stimulates both the release and infectivity of HIV-1 viral particles by an editing-dependent mechanism where it associates with viral RNAs and edits adenosines in the 5'UTR and the Rev and Tat coding sequence. Can enhance viral replication of HDV via A-to-I editing at a site designated as amber/W, thereby changing an UAG amber stop codon to an UIG tryptophan (W) codon that permits synthesis of the large delta antigen (L-HDAg) which has a key role in the assembly of viral particles. However, high levels of ADAR1 inhibit HDV replication. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAR
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1066
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene Adar
Gene Accession No. P55265
Gene Alias 136 kDa double-stranded RNA-binding protein; ADA; Adar; ADAR1; Adar1p110; Adar1p150; Adenosine Deaminase; adenosine deaminase acting on RNA 1-A; adenosine deaminase RNA specific; adenosine deaminase, RNA specific; adenosine deaminase, RNA-specific; AGS6; AV242451; double-stranded RNA-specific adenosine deaminase; DRADA; DSH; DSRAD; dsRNA adenosine deaminase; dsRNA adeonosine deaminase; G1P1; IFI4; IFI-4; interferon-induced protein 4; interferon-inducible protein 4; K88DSRBP; LOW QUALITY PROTEIN: double-stranded RNA-specific adenosine deaminase; mZaADAR; P136; RNA adenosine deaminase 1; RNA-specific adenosine deaminase p110 form; RNA-specific adenosine deaminase p150 form
Gene Symbols Adar
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human ADAR1. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.