Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584043 Supplier Invitrogen™ Supplier No. PA584043
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDYLTSLKSV PYGSEEYLQL RSKVHLRSAR RLCELCCANR GTFIKVGQHL GALDYLLPEE YTSTLKVLHS QAPQSSMQEI RQVIRGDLGK E Highest antigen sequence indentity to the following orthologs: Rat - 30%, Mouse - 93%.

ADCK1 is a protein coding gene. The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADCK1
Gene Accession No. Q86TW2
Gene Alias 2610005A10Rik; aarF domain containing kinase 1; AarF domain-containing protein kinase 1; Adck1; Nrbf1; uncharacterized aarF domain-containing protein kinase 1
Gene Symbols ADCK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADCK1. Recombinant protein control fragment (Product #RP-100534).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.