Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCY10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114825 Supplier Invitrogen™ Supplier No. PA5114825
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADCY10.

SAC belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction.The protein encoded by this gene belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCY10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ADCY10
Gene Accession No. Q8C0T9, Q96PN6, Q9Z286
Gene Alias 3',5'-cyclic AMP synthetase; 4930431D04Rik; 4931412F17; Adcy10; adenylate cyclase 10; adenylate cyclase 10 (soluble); adenylate cyclase 10, soluble; Adenylate cyclase homolog; Adenylate cyclase type 10; AH-related protein; ATP pyrophosphate-lyase; epididymis secretory sperm binding protein Li 7a; germ cell soluble adenylyl cyclase; HCA2; HEL-S-7a; hsAC; RP1-313L4.2; Sac; SACI; Sacy; soluble adenylyl cyclase; testicular soluble adenylyl cyclase; testicular soluble adenylyl cyclase (SAC)
Gene Symbols ADCY10
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADCY10(Accession Q96PN6), corresponding to amino acid residues F29-I79.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271639, 55811, 59320
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.