Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenine 991KG
Supplier: Sigma Aldrich Fine Chemicals Biosciences A8626
Adenine is a purine nucleobase. It is part of DNA and RNA. Adenine is also a component of cofactors (NAD FAD) and signaling molecules (cAMP).