Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenine Hydrochloride, ∼99%, MP Biomedicals™

Catalog No. 0210019125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-100-19125 25 g
ICN10019105 5 g
ICN10019180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-100-19125 Supplier MP Biomedicals, Inc Supplier No. 0210019125
Add to Cart
Add to Cart

Adenine Hydrochloride, ∼99% is used in viral research and has a wide range of chemical and biochemical roles in vivo and in vitro.

  • Adenine has a wide range of chemical and biochemical roles in vivo and in vitro.
  • It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).
  • It is used as a blood preservative and in viral research.
  • It is a purine nucleobase widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids.
  • It is a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-72-7
Molecular Formula C5H8ClN5O
Molecular Weight (g/mol) 189.60
MDL Number MFCD11046306
InChI Key MYRDTAUFFBYTHA-UHFFFAOYSA-N
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
PubChem CID 24211900
IUPAC Name 7H-purin-6-amine hydrate hydrochloride
SMILES O.Cl.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance λ max 262 nm (0.1 N HCl)
Color White
Assay Percent Range 0.992
Quantity 25 g
Formula Weight 171.6
Percent Purity 99%
Physical Form Powder
For Use With (Application) Used as a blood preservative and in viral research, Regulatory molecule
Chemical Name or Material Adenine hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.