Learn More
Clontech Labs 3P ADENO-X VIRUS PURIFICATION KIT
SDP
Supplier:  Clontech Labs 3P 631532
Encompass
BD Adeno-X Virus Purification Kit, 1 Preparation, The BD Adeno-X Virus Purification Kit is a complete chromatography-based system that enables you to easily and rapidly purify amplified adenovirus, Each kit has wide range of viral capacity
Catalog No. NC9996987
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.