Learn More
Tocris Bioscience™ Adenosine 5'-(a,b-methylene)diphosphate sodium salt

Description
Specifications
Specifications
| Purity | >98% |
| Gene Alias | AMPCP, APCP |
| Quantity | 10 mg |
| Product Type | Adenosine 5'-(α,β-methylene)diphosphate sodium salt |
| Molecular Weight (g/mol) | 447.21 |
| Inhibitors | Adenosine 5'-(a,b-methylene)diphosphate sodium salt |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.