Learn More
Medchemexpress LLC Adenosine, 5'-deoxy-5'-fluoro | 731-98-6 | N/A | 96.2% | 269.23 | C10H12FN5O3 | 100 MG

Supplier: Medchemexpress LLC HYW616732100MG
5′-Fluoro-5′-deoxyadenosine is a fluorinated adenosine nucleoside provided for research use in biochemical and pharmacological studies. The manufacturer supplies solubility guidance and recommended storage conditions for both solid and solution forms.
- Fluorinated adenosine nucleoside suitable for research use.
- Purity 96.2% (typical, by HPLC).
- Molecular weight 269.23 g/mol.
- Soluble in DMSO (≈20 mg/mL) with suggested in vivo formulation protocols.
- Recommended storage: solid at 4°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.