Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-diphophate monopotassium salt, 98 to 100%, MP Biomedicals™

Catalog No. ICN15025901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15025901 1 g
ICN15025905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15025901 Supplier MP Biomedicals, Inc Supplier No. 0215025901
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 98 to 100%
Quantity 1 g
IUPAC Name potassium;(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolan-3-olate;dihydrate
Formula Weight 465.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphophate monopotassium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.