Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals™

Catalog No. MP210005605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210005605 5 g
MP21000565 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210005605 Supplier MP Biomedicals, Inc Supplier No. 0210005605
Add to Cart
Add to Cart

Involved in energy storage and nucleic acid metabolism. Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals is an adenine nucleotide that converts into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors.

ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Quantity 5 g
Optical Rotation [α]20/D -29.6±1° c = 0.5% in 0.5M Na2HPO4 (Lit.)
Formula Weight 471.2
Percent Purity ≥95%
Physical Form Powder
Recommended Storage Store at −20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.