Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Diphosphate Monopotassium Salt Dihydrate, MP Biomedicals™

Catalog No. MP021502595 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021502595 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021502595 Supplier MP Biomedicals, Inc Supplier No. 02150259.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acts as a P2Y receptor agonist. Adenosine-5'-Diphosphate Monopotassium Salt Dihydrate, MP Biomedicals has been used to study the release, neuronal effects and removal of extracellular β-NAD+ and to compare prasugrel versus clopidogrel antiplatelet th

Adenosine 5′-diphosphate has been used to study the release, neuronal effects and removal of extracellular β-nicotinamide adenine dinucleotide (β-NAD+) in the rat brain. Adenosine 5′-diphosphate has also been used to compare prasugrel versus clopidogrel antiplatelet therapy after acute coronary syndrome.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
IUPAC Name potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Quantity 500 mg
Solubility Information Soluble in water (5mg/mL, clear and colorless solution).
Formula Weight 501.2
Percent Purity >98%
Grade Not Available
Physical Form Powder
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.