Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Triphosphate Trihydrate, MP Biomedicals™

Catalog No. ICN19461301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19461301 1 g
ICN19461305 5 g
ICN19461310 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19461301 Supplier MP Biomedicals, Inc Supplier No. 0219461301
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51963-61-2
Molecular Formula C10H20N5Na2O16P3
Molecular Weight (g/mol) 605.19
MDL Number MFCD00150754
InChI Key MWEQTWJABOLLOS-MVHBFMHHNA-L
Synonym adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
PubChem CID 134129625
IUPAC Name disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
SMILES O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Physical Form Solid
Quantity 1 g
Formula Weight 605.23
Chemical Name or Material Adenosine-5'-Triphosphate Trihydrate
Recommended Storage Store at 0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.