Learn More
Medchemexpress LLC HY-128064 5mg Medchemexpress, Adenosine amine congener CAS:96760-69-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1280645MG
Encompass_Preferred
Medchemexpress, HY-128064 5mg Adenosine amine congener CAS:96760-69-9 Adenosine amine congener (ADAC) is a selective A1 adenosine receptor agonist, can ameliorate noise- and Cisplatin-induced cochlear injury. Adenosine amine congener also has neuroprotective effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.