Learn More
Selleck Chemical LLC Adenosine Dialdehyde (ADOX) 5mg 34240-05-6
SDP
Supplier:  Selleck Chemical LLC S86085MG
Encompass_Preferred
Adenosine Dialdehyde (ADOX) is an adenosine analog and S-adenosylmethionine-dependent methyltransferase inhibitor with an IC50 of 40 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.