Learn More
Cayman Chemical Adenosine 5'-(γ-thio)-triphosphate (lithium salt), 93839-89-5, 5 mg
SDP
Supplier:  Cayman Chemical 149575
Encompass_Preferred
A stable analog of ATP that acts as a potent agonist of G protein-coupled P2Y2 and P2Y11 receptors (pEC50 = 5.52 for P2Y11)
Catalog No. NC1399190
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.