Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Cyclase 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555041 Supplier Invitrogen™ Supplier No. PA555041
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRRNSGATFT EGSWSPELPF DNIVGKQNTL AALTRNSINL LPNHLAQALH VQSGPEEINK RIEHTIDLRS GDKLRREHIK PFSLMF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Adcy8
Gene Accession No. P40145
Gene Alias AC8; ADCY3; ADCY8; adenylate cyclase 8; adenylate cyclase 8 (brain); adenylate cyclase type 8; Adenylate cyclase type VIII; adenylyl cyclase 8; adenylyl cyclase-8, brain; ATP pyrophosphate-lyase 8; AW060868; ca(2+)/calmodulin-activated adenylyl cyclase; epididymis secretory sperm binding protein Li 172mP; HBAC1; HEL-S-172mP; type VIII adenylyl cyclase
Gene Symbols Adcy8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Adenylate Cyclase 8. Recombinant protein control fragment (Product #RP-93838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.