Learn More
CTMATTER LLC Adesso NuScan 2500TU Spill Resistant Antimicrobial 2D Barcode Scanner
Supplier:  CTMATTER LLC NUSCAN2500TU
Encompass_Preferred
This NuScan 2500TU Spill Resistant Antimicrobial 2D Barcode Scanner from Adesso is a reliable and efficient plug and play scanner. The scanner is compatible with major 2D barcodes including PDF 417, QR Codes, Micro QR Codes, and Data Matrix, and for 1D barcodes the unit is compatible with UPC-A, Code 39, Code 128, and more.
Catalog No. NC3861286